Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCGAGGGTGCAGAGGGAGGTCA[C/T]GACTTTCAGTCTGCTGTTTAGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601691 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA4 - ATP binding cassette subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000350.2 | 6871 | Missense Mutation | CAT,CGT | H,R 2256 | NP_000341.2 |