Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCTCGGGCAGCGACTCCTCCGA[G/T]TCGGTGCGCAGGTCCTCGTCGTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609895 MIM: 601791 | ||||||||||||||||||||
Literature Links: |
CASZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CASZ1 - castor zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079843.2 | 5774 | Missense Mutation | GAA,GAC | E,D 1718 | NP_001073312.1 | |
NM_017766.4 | 5774 | Intron | NP_060236.3 | |||
XM_005263479.3 | 5774 | Missense Mutation | GAA,GAC | E,D 1742 | XP_005263536.1 | |
XM_011541635.2 | 5774 | Intron | XP_011539937.1 | |||
XM_017001539.1 | 5774 | Missense Mutation | GAA,GAC | E,D 1842 | XP_016857028.1 | |
XM_017001540.1 | 5774 | Missense Mutation | GAA,GAC | E,D 1818 | XP_016857029.1 | |
XM_017001541.1 | 5774 | Missense Mutation | GAA,GAC | E,D 1818 | XP_016857030.1 |
PEX14 - peroxisomal biogenesis factor 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |