Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGCAATAACTAACACCGTTCTT[C/T]CATCCATCAGTCGATCTAGAGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB10 - ATP binding cassette subfamily B member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012089.2 | 2859 | Intron | NP_036221.2 | |||
XM_011544135.2 | 2859 | Missense Mutation | GAA,GGA | E,G 503 | XP_011542437.1 | |
XM_011544136.2 | 2859 | Intron | XP_011542438.1 |
LOC101927478 - uncharacterized LOC101927478 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |