Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCGGGCTTCCGCAGCTGCCTGCC[C/G]GGCGTGAGCCAGCTCCTTCGGCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614269 MIM: 609971 MIM: 605851 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR153 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR153 - G protein-coupled receptor 153 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HES3 - hes family bHLH transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024598.3 | 287 | Silent Mutation | CCC,CCG | P,P 74 | NP_001019769.1 |
ICMT - isoprenylcysteine carboxyl methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC00337 - long intergenic non-protein coding RNA 337 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |