Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCTGGTCCACCAGACTGAACTC[A/G]CGGATGAAGTAGTAGAAGTGCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKNK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKNK1 - MAP kinase interacting serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOB3C - MOB kinase activator 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145279.4 | 3744 | Silent Mutation | CGC,CGT | R,R 246 | NP_660322.2 | |
NM_201403.2 | 3744 | Silent Mutation | CGC,CGT | R,R 194 | NP_958805.1 | |
XM_017000407.1 | 3744 | Silent Mutation | CGC,CGT | R,R 217 | XP_016855896.1 |