Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGCATTGTAGGAGCTCTGCTG[A/C]ATGTGGCTGCCCCCGAACATGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBX15 - T-box 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152380.2 | 1956 | Missense Mutation | ATG,ATT | M,I 395 | NP_689593.2 | |
XM_005271161.3 | 1956 | Missense Mutation | ATG,ATT | M,I 534 | XP_005271218.1 | |
XM_005271162.1 | 1956 | Missense Mutation | ATG,ATT | M,I 501 | XP_005271219.1 | |
XM_011542052.2 | 1956 | Missense Mutation | ATG,ATT | M,I 219 | XP_011540354.1 |