Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGCTAAGGTTGCACTGGAAGAT[G/T]TTCTAGCAGCTTTCTCTGATTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HHIPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HHIPL2 - HHIP like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024746.3 | 1997 | Missense Mutation | AAA,ACA | K,T 646 | NP_079022.2 | |
XM_011509985.2 | 1997 | Missense Mutation | AAA,ACA | K,T 665 | XP_011508287.1 | |
XM_011509986.1 | 1997 | Intron | XP_011508288.1 | |||
XM_017002350.1 | 1997 | Intron | XP_016857839.1 |