Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGTAGAGACCTGGACCCCCGGAA[A/T]GGTGGCAACTTGGCTGAGAGGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603272 MIM: 616698 | ||||||||||||||||||||
Literature Links: |
CNKSR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNKSR1 - connector enhancer of kinase suppressor of Ras 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297647.1 | 90 | Missense Mutation | AAG,ATG | K,M 11 | NP_001284576.1 | |
NM_001297648.1 | 90 | UTR 5 | NP_001284577.1 | |||
NM_006314.2 | 90 | Missense Mutation | AAG,ATG | K,M 11 | NP_006305.2 | |
XM_011540484.2 | 90 | UTR 5 | XP_011538786.1 | |||
XM_011540485.2 | 90 | Intron | XP_011538787.1 |
ZNF593 - zinc finger protein 593 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |