Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACTCTGCAATCCTTCTGAACC[A/G]CCTCCACCCAGAAGACTAAATGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603497 | ||||||||||||||||||||
Literature Links: |
DYRK3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYRK3 - dual specificity tyrosine phosphorylation regulated kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004023.1 | 430 | Silent Mutation | CCA,CCG | P,P 36 | NP_001004023.1 | |
NM_003582.2 | 430 | Silent Mutation | CCA,CCG | P,P 56 | NP_003573.2 | |
XM_005273315.4 | 430 | Silent Mutation | CCA,CCG | P,P 36 | XP_005273372.1 | |
XM_011510061.2 | 430 | Silent Mutation | CCA,CCG | P,P 21 | XP_011508363.1 |