Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTCTTCGAGGGCTTCTACGGTGT[A/C]ACCTTGCTGCACATCGACTGGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609144 MIM: 610864 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLVCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLVCR1 - feline leukemia virus subgroup C cellular receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014053.3 | 597 | Silent Mutation | GTA,GTC | V,V 141 | NP_054772.1 | |
XM_011509446.2 | 597 | Silent Mutation | GTA,GTC | V,V 141 | XP_011507748.1 | |
XM_011509447.2 | 597 | Silent Mutation | GTA,GTC | V,V 141 | XP_011507749.1 | |
XM_011509448.1 | 597 | Silent Mutation | GTA,GTC | V,V 141 | XP_011507750.1 |
FLVCR1-AS1 - FLVCR1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |