Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTCTTCTGTGCAGTGCAAAAGA[C/T]ATGAAACATCGGCTAGGTTTCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602516 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS4 - regulator of G-protein signaling 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102445.2 | 674 | Silent Mutation | GAC,GAT | D,D 115 | NP_001095915.1 | |
NM_001113380.1 | 674 | UTR 5 | NP_001106851.1 | |||
NM_001113381.1 | 674 | Silent Mutation | GAC,GAT | D,D 18 | NP_001106852.1 | |
NM_005613.5 | 674 | Silent Mutation | GAC,GAT | D,D 18 | NP_005604.1 |