Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGTAGGAATTGGTGAAGCCAAC[A/G]TAGTCATAGCCAGAGAGATGGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR10Z1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR10Z1 - olfactory receptor family 10 subfamily Z member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPTA1 - spectrin alpha, erythrocytic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003126.2 | 7322 | Silent Mutation | TAC,TAT | Y,Y 2409 | NP_003117.2 | |
XM_011509916.2 | 7322 | Silent Mutation | TAC,TAT | Y,Y 2409 | XP_011508218.1 | |
XM_011509917.2 | 7322 | Silent Mutation | TAC,TAT | Y,Y 2403 | XP_011508219.1 | |
XM_011509918.2 | 7322 | Intron | XP_011508220.1 | |||
XM_011509919.2 | 7322 | Intron | XP_011508221.1 |