Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCACCTCTCTCCCGCAGAACGGC[A/G]ACTACAACAAGCCGATCCCCGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM201 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM201 - transmembrane protein 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010866.3 | 298 | Missense Mutation | AAC,GAC | N,D 81 | NP_001010866.1 | |
NM_001130924.2 | 298 | Missense Mutation | AAC,GAC | N,D 81 | NP_001124396.2 | |
XM_006710417.3 | 298 | Missense Mutation | AAC,GAC | N,D 81 | XP_006710480.1 | |
XM_011540910.2 | 298 | Missense Mutation | AAC,GAC | N,D 81 | XP_011539212.1 | |
XM_017000549.1 | 298 | Missense Mutation | AAC,GAC | N,D 81 | XP_016856038.1 | |
XM_017000550.1 | 298 | Missense Mutation | AAC,GAC | N,D 81 | XP_016856039.1 | |
XM_017000551.1 | 298 | Missense Mutation | AAC,GAC | N,D 43 | XP_016856040.1 | |
XM_017000552.1 | 298 | Missense Mutation | AAC,GAC | N,D 43 | XP_016856041.1 |