Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGCTGGTGTAGAACTGCAGCTCC[A/G]GGGCCCTGTGGGTGACAGAACACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK11B - cyclin dependent kinase 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35E2B - solute carrier family 35 member E2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110781.2 | 1688 | Missense Mutation | CCG,CTG | P,L 256 | NP_001104251.1 | |
NM_001290264.1 | 1688 | Missense Mutation | CCG,CTG | P,L 256 | NP_001277193.1 | |
XM_011542072.1 | 1688 | Intron | XP_011540374.1 |