Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGGAAAGCGGCTTGGACCACG[A/T]TGGGGTGGAGGTGCTGGAAGATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609794 MIM: 602943 | ||||||||||||||||||||
Literature Links: |
LINGO4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINGO4 - leucine rich repeat and Ig domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RORC - RAR related orphan receptor C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001523.1 | 2366 | Missense Mutation | AAC,ATC | N,I 471 | NP_001001523.1 | |
NM_005060.3 | 2366 | Missense Mutation | AAC,ATC | N,I 492 | NP_005051.2 | |
XM_006711484.3 | 2366 | Missense Mutation | AAC,ATC | N,I 625 | XP_006711547.2 |