Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCTTGGACAGCCCGGGCAACCT[C/T]GACACCCTGCAGGCGAAAAAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRRX1 - paired related homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006902.4 | 122 | Silent Mutation | CTC,CTT | L,L 25 | NP_008833.1 | |
NM_022716.3 | 122 | Silent Mutation | CTC,CTT | L,L 25 | NP_073207.1 | |
XM_006711388.3 | 122 | Intron | XP_006711451.1 |