Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTGAATTAACACAGTCAAGAGC[A/G]AAAGTATGTATCTTGAAATAAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616876 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC18 - coiled-coil domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306076.1 | 1104 | Silent Mutation | GCA,GCG | A,A 153 | NP_001293005.1 | |
NM_206886.4 | 1104 | Silent Mutation | GCA,GCG | A,A 153 | NP_996769.3 | |
XM_011541361.2 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_011539663.2 | |
XM_011541372.2 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_011539674.2 | |
XM_017001154.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856643.1 | |
XM_017001155.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856644.1 | |
XM_017001156.1 | 1104 | Silent Mutation | GCA,GCG | A,A 261 | XP_016856645.1 | |
XM_017001157.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856646.1 | |
XM_017001158.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856647.1 | |
XM_017001159.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856648.1 | |
XM_017001160.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856649.1 | |
XM_017001161.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856650.1 | |
XM_017001162.1 | 1104 | Silent Mutation | GCA,GCG | A,A 153 | XP_016856651.1 | |
XM_017001163.1 | 1104 | Silent Mutation | GCA,GCG | A,A 153 | XP_016856652.1 | |
XM_017001164.1 | 1104 | Silent Mutation | GCA,GCG | A,A 153 | XP_016856653.1 | |
XM_017001165.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856654.1 | |
XM_017001166.1 | 1104 | Silent Mutation | GCA,GCG | A,A 64 | XP_016856655.1 | |
XM_017001167.1 | 1104 | Silent Mutation | GCA,GCG | A,A 52 | XP_016856656.1 | |
XM_017001168.1 | 1104 | UTR 5 | XP_016856657.1 | |||
XM_017001169.1 | 1104 | Silent Mutation | GCA,GCG | A,A 271 | XP_016856658.1 | |
XM_017001170.1 | 1104 | Intron | XP_016856659.1 | |||
XM_017001171.1 | 1104 | Intron | XP_016856660.1 |
TMED5 - transmembrane p24 trafficking protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |