Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCACCACCTCCTTTACCTGTCC[C/G]AAAGTCAATGGCTCTCAGGGCGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM63 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM63 - tripartite motif containing 63 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032588.3 | 1113 | Missense Mutation | CGG,GGG | R,G 325 | NP_115977.2 | |
XM_017002559.1 | 1113 | Missense Mutation | CGG,GGG | R,G 325 | XP_016858048.1 |