Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTACCTATATGAGGATTGAGAGC[C/T]GTCACCAACATTAGAGACTCATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 136850 | ||||||||||||||||||||
Literature Links: |
FH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FH - fumarate hydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000143.3 | 1874 | Silent Mutation | ACA,ACG | T,T 457 | NP_000134.2 | |
XM_011544132.2 | 1874 | Silent Mutation | ACA,ACG | T,T 381 | XP_011542434.1 |