Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACTGGCTACCTGTCCAACAACC[A/G]CTTCTATCCACCGCCGCCCAAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606210 | ||||||||||||||||||||
Literature Links: |
SEPN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SEPN1 - selenoprotein N, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020451.2 | 771 | Missense Mutation | CAC,CGC | H,R 239 | NP_065184.2 | |
NM_206926.1 | 771 | Missense Mutation | CAC,CGC | H,R 205 | NP_996809.1 |