Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCCATAATCCTGAGCAATGGTG[C/T]GCTTCGGGTCTGATACCAAAGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609831 MIM: 176763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMACHC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMACHC - methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDX1 - peroxiredoxin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202431.1 | 670 | Missense Mutation | CAC,CGC | H,R 110 | NP_001189360.1 | |
NM_002574.3 | 670 | Missense Mutation | CAC,CGC | H,R 110 | NP_002565.1 | |
NM_181696.2 | 670 | Missense Mutation | CAC,CGC | H,R 110 | NP_859047.1 | |
NM_181697.2 | 670 | Intron | NP_859048.1 |