Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTACAGCAGAAGTAGAGAATCC[A/G]AGGATTTTGGAAGAAGAAATTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610656 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KYAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KYAT3 - kynurenine aminotransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008661.2 | 458 | Silent Mutation | CTC,CTT | L,L 27 | NP_001008661.1 | |
NM_001008662.2 | 458 | Intron | NP_001008662.1 |
RBMXL1 - RNA binding motif protein, X-linked like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162536.2 | 458 | UTR 5 | NP_001156008.1 | |||
NM_019610.5 | 458 | Intron | NP_062556.2 |