Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAGCACACAGTCAGTCTACATC[A/C]AGGTGAGTCCCTTGGCCAGCGTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139392 MIM: 601271 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GUCA2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GUCA2A - guanylate cyclase activator 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GUCA2B - guanylate cyclase activator 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007102.2 | 118 | Missense Mutation | AAG,CAG | K,Q 30 | NP_009033.1 |