Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCTGTTACCTGGTGAAGGAGC[A/G]GGGCTTGGCTGCAGGAGATGCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP23 - dual specificity phosphatase 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319658.1 | 391 | Missense Mutation | CAG,CGG | Q,R 114 | NP_001306587.1 | |
NM_001319659.1 | 391 | Missense Mutation | CAG,CGG | Q,R 114 | NP_001306588.1 | |
NM_017823.4 | 391 | Missense Mutation | CAG,CGG | Q,R 114 | NP_060293.2 |