Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTATCGCACAGCAGAGGACCATGC[A/T]GAATATCATGCCAAAGATCTGAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613133 | ||||||||||||||||||||
Literature Links: |
TSPAN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPAN2 - tetraspanin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308315.1 | 612 | Missense Mutation | AGC,TGC | S,C 182 | NP_001295244.1 | |
NM_001308316.1 | 612 | Missense Mutation | AGC,TGC | S,C 179 | NP_001295245.1 | |
NM_005725.5 | 612 | Missense Mutation | AGC,TGC | S,C 207 | NP_005716.2 | |
XM_016999996.1 | 612 | Missense Mutation | AGC,TGC | S,C 154 | XP_016855485.1 |