Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCCACCTATCCTGCCCCCTATCT[C/G]TCCCTGAACAATGGCTTCCTGGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606578 MIM: 605867 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP10 - aquaporin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080429.2 | 502 | Silent Mutation | CTC,CTG | L,L 154 | NP_536354.2 | |
XM_011510104.2 | 502 | Silent Mutation | CTC,CTG | L,L 155 | XP_011508406.1 |
ATP8B2 - ATPase phospholipid transporting 8B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |