Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCAGTCAGCAGCTGAGCCATCCT[C/T]CAGCTCTGAAAGCTCAGTCTTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 164873 | ||||||||||||||||||||
Literature Links: |
CYCSP52 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYCSP52 - cytochrome c, somatic pseudogene 52 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETV3 - ETS variant 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145312.1 | 872 | Missense Mutation | AAG,GAG | K,E 195 | NP_001138784.1 | |
NM_005240.1 | 872 | Intron | NP_005231.1 | |||
XM_006711210.2 | 872 | Missense Mutation | AAG,GAG | K,E 195 | XP_006711273.1 |