Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGAATGGGAAGGTGACGAATGA[A/G]GGGACCACATCTACGCTGACAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VCAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VCAM1 - vascular cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078.3 | 443 | Silent Mutation | GAA,GAG | E,E 74 | NP_001069.1 | |
NM_001199834.1 | 443 | Intron | NP_001186763.1 | |||
NM_080682.2 | 443 | Silent Mutation | GAA,GAG | E,E 74 | NP_542413.1 |