Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTCAAACGTATGTGTTCTTGGC[C/T]CTTGGTGGAACTGAAGCCCTTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR2AK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR2AK2 - olfactory receptor family 2 subfamily AK member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004491.1 | 363 | Silent Mutation | GCC,GCT | A,A 121 | NP_001004491.1 |
OR2L13 - olfactory receptor family 2 subfamily L member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304535.1 | 363 | Intron | NP_001291464.1 | |||
NM_175911.3 | 363 | Intron | NP_787107.1 | |||
XM_011544169.2 | 363 | Intron | XP_011542471.1 |