Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCCGGAGAATGAAGAAGAGGGT[A/G]GCACATGTGGCAAAGCCAAAAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612037 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUL1 - mitochondrial E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024544.2 | 1599 | Silent Mutation | GCC,GCT | A,A 253 | NP_078820.2 | |
XM_011542137.2 | 1599 | Silent Mutation | GCC,GCT | A,A 214 | XP_011540439.1 |