Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACTCCTTTATTCATTATGCCC[A/G]TCTTCTCCTGAACTCCTAAAATCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 114230 MIM: 602143 | ||||||||||||||||||||
Literature Links: |
CAPN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN2 - calpain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TP53BP2 - tumor protein p53 binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031685.2 | 3471 | Missense Mutation | ACG,ATG | T,M 1060 | NP_001026855.2 | |
NM_005426.2 | 3471 | Missense Mutation | ACG,ATG | T,M 931 | NP_005417.1 | |
XM_011544268.1 | 3471 | Missense Mutation | ACG,ATG | T,M 993 | XP_011542570.1 | |
XM_011544269.2 | 3471 | Missense Mutation | ACG,ATG | T,M 931 | XP_011542571.1 | |
XM_017002220.1 | 3471 | Missense Mutation | ACG,ATG | T,M 993 | XP_016857709.1 |