Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGCTGATTCAGGGGGGTCAAT[C/G]CTCCTCTAGAGAAGAGCCACACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605593 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTMR11 - myotubularin related protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145862.1 | 2492 | Missense Mutation | GCA,GGA | A,G 568 | NP_001139334.1 | |
NM_181873.3 | 2492 | Missense Mutation | GCA,GGA | A,G 496 | NP_870988.2 | |
XM_006711137.1 | 2492 | Missense Mutation | GCA,GGA | A,G 496 | XP_006711200.1 | |
XM_011509099.2 | 2492 | UTR 3 | XP_011507401.1 |
SF3B4 - splicing factor 3b subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |