Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTGCGGCTGTGTCAGATTTCT[A/G]TGTTCCTGTCTCTGAAATGCCTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609853 | ||||||||||||||||||||
Literature Links: |
PPCS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPCS - phosphopantothenoylcysteine synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077447.2 | 205 | Missense Mutation | TAT,TGT | Y,C 12 | NP_001070915.1 | |
NM_001287506.1 | 205 | Missense Mutation | TAT,TGT | Y,C 12 | NP_001274435.1 | |
NM_001287507.1 | 205 | UTR 5 | NP_001274436.1 | |||
NM_001287508.1 | 205 | Missense Mutation | TAT,TGT | Y,C 12 | NP_001274437.1 | |
NM_001287509.1 | 205 | Missense Mutation | TAT,TGT | Y,C 12 | NP_001274438.1 | |
NM_001287510.1 | 205 | Missense Mutation | TAT,TGT | Y,C 12 | NP_001274439.1 | |
NM_001287511.1 | 205 | Missense Mutation | TAT,TGT | Y,C 185 | NP_001274440.1 | |
NM_024664.3 | 205 | Missense Mutation | TAT,TGT | Y,C 185 | NP_078940.2 |
ZMYND12 - zinc finger MYND-type containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |