Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGCTGCCCAGCCCAGGGCAGCC[A/G]TGCCAAGTATGTTCTTAGCAGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607622 | ||||||||||||||||||||
Literature Links: |
PBXIP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PBXIP1 - PBX homeobox interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317734.1 | 1584 | Missense Mutation | CGG,TGG | R,W 596 | NP_001304663.1 | |
NM_001317735.1 | 1584 | Missense Mutation | CGG,TGG | R,W 470 | NP_001304664.1 | |
NM_020524.3 | 1584 | Missense Mutation | CGG,TGG | R,W 625 | NP_065385.2 |
PMVK - phosphomevalonate kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |