Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGGGACAGGGGCTGCCGGAGG[C/G]CTTGCCGTAGGTGGCTTCGAGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC5A9 - solute carrier family 5 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011547.2 | 352 | Silent Mutation | GGC,GGG | G,G 104 | NP_001011547.2 | |
NM_001135181.1 | 352 | Silent Mutation | GGC,GGG | G,G 104 | NP_001128653.1 | |
XM_011540924.2 | 352 | Silent Mutation | GGC,GGG | G,G 104 | XP_011539226.1 | |
XM_011540925.2 | 352 | Silent Mutation | GGC,GGG | G,G 104 | XP_011539227.1 | |
XM_011540926.2 | 352 | UTR 5 | XP_011539228.1 | |||
XM_011540927.1 | 352 | UTR 5 | XP_011539229.1 | |||
XM_011540928.1 | 352 | UTR 5 | XP_011539230.1 | |||
XM_011540929.2 | 352 | Silent Mutation | GGC,GGG | G,G 104 | XP_011539231.1 | |
XM_017000558.1 | 352 | Silent Mutation | GGC,GGG | G,G 104 | XP_016856047.1 |