Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTAGGACTCACCTGAGCAGATG[C/A]CAGCCACATCTTCCTGGTGGGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602592 | ||||||||||||||||||||
Literature Links: |
CD5L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD5L - CD5 molecule like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005894.2 | 1171 | Missense Mutation | GGC,GTC | G,V 343 | NP_005885.1 | |
XM_005245602.1 | 1171 | Missense Mutation | GGC,GTC | G,V 343 | XP_005245659.1 | |
XM_017002806.1 | 1171 | Missense Mutation | GGC,GTC | G,V 343 | XP_016858295.1 |