Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCGAGATGCTTCCCATCAGATC[C/T]GCCAGGAGGCCCGGGTGCTCCGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610624 MIM: 608953 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADPRHL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADPRHL2 - ADP-ribosylhydrolase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEKT2 - tektin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014466.2 | 214 | Missense Mutation | CGC,TGC | R,C 39 | NP_055281.2 | |
XM_005270753.2 | 214 | Missense Mutation | CGC,TGC | R,C 39 | XP_005270810.1 | |
XM_011541258.2 | 214 | Missense Mutation | CGC,TGC | R,C 39 | XP_011539560.1 | |
XM_017001055.1 | 214 | Missense Mutation | CGC,TGC | R,C 39 | XP_016856544.1 |