Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCTCTTGGGGCCCCCACCCAGG[C/T]CAGGATGCGCCTGGGGCATAAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHDC1 - AT-hook DNA binding motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029882.3 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | NP_001025053.1 | |
XM_005245848.3 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_005245905.1 | |
XM_005245849.3 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_005245906.1 | |
XM_005245850.3 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_005245907.1 | |
XM_005245851.3 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_005245908.1 | |
XM_005245852.3 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_005245909.1 | |
XM_011541255.1 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_011539557.1 | |
XM_011541256.2 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_011539558.1 | |
XM_011541257.2 | 5790 | Missense Mutation | GAC,GGC | D,G 1576 | XP_011539559.1 |