Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAGACTGCCAGAAGAGGCTGAA[G/T]AACTGCCAAAGTCATCACTATACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611578 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRRS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRRS1 - ferric chelate reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013660.2 | 1504 | Missense Mutation | ATT,CTT | I,L 459 | NP_001013682.2 | |
XM_005270861.3 | 1504 | Missense Mutation | ATT,CTT | I,L 459 | XP_005270918.1 | |
XM_011541451.2 | 1504 | Missense Mutation | ATT,CTT | I,L 459 | XP_011539753.1 | |
XM_011541452.1 | 1504 | Missense Mutation | ATT,CTT | I,L 459 | XP_011539754.1 | |
XM_017001271.1 | 1504 | Missense Mutation | ATT,CTT | I,L 459 | XP_016856760.1 |