Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTGCCTGAGAATCTCCTTCTT[G/T]CTAGGTGGAATTGAATCACTGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724919 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724919 - uncharacterized LOC102724919 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDC - phosducin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002597.4 | 209 | Missense Mutation | AGA,AGC | R,S 45 | NP_002588.3 | |
NM_022576.3 | 209 | UTR 5 | NP_072098.1 | |||
XM_011509603.2 | 209 | Missense Mutation | AGA,AGC | R,S 45 | XP_011507905.1 |