Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGCTGCAGGCAGGACTCCAGC[A/G]TCAGGATGTTGCCACTTGGAGTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACOT11 - acyl-CoA thioesterase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015547.3 | 1389 | Intron | NP_056362.1 | |||
NM_147161.3 | 1389 | UTR 3 | NP_671517.1 |
FAM151A - family with sequence similarity 151 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176782.2 | 1389 | Intron | NP_788954.2 | |||
XM_006710599.3 | 1389 | Missense Mutation | ACG,ATG | T,M 368 | XP_006710662.1 |