Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGACAGCCACCTGCTCCGGTAT[A/C]TCTCCTGGACTGTGTACCCTGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCNKB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCNKB - chloride voltage-gated channel Kb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000085.4 | 432 | Missense Mutation | ATC,CTC | I,L 94 | NP_000076.2 | |
NM_001165945.2 | 432 | Intron | NP_001159417.2 |