Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGACATTCCAGGATGTGGCTG[G/T]GGATTTCACTGCAGAGGAGTGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF684 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF684 - zinc finger protein 684 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152373.3 | 628 | Missense Mutation | GGG,GTG | G,V 15 | NP_689586.3 | |
XM_011540671.2 | 628 | Missense Mutation | GGG,GTG | G,V 15 | XP_011538973.1 | |
XM_011540672.2 | 628 | Missense Mutation | GGG,GTG | G,V 15 | XP_011538974.1 | |
XM_011540673.2 | 628 | Missense Mutation | GGG,GTG | G,V 15 | XP_011538975.1 | |
XM_017000290.1 | 628 | Missense Mutation | GGG,GTG | G,V 15 | XP_016855779.1 | |
XM_017000291.1 | 628 | UTR 5 | XP_016855780.1 | |||
XM_017000292.1 | 628 | Intron | XP_016855781.1 |