Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTCCTGGCAGCCCTGCTGCTG[A/G]GGGCGACAGGCACAGGGGTCGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609034 | ||||||||||||||||||||
Literature Links: |
HEYL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEYL - hes related family bHLH transcription factor with YRPW motif-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014571.3 | 836 | Missense Mutation | CCC,CTC | P,L 262 | NP_055386.1 | |
XM_005270745.2 | 836 | Missense Mutation | CCC,CTC | P,L 234 | XP_005270802.1 |