Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAAAAAACGGTTTTTAACCCAT[C/T]GGTTTTGTTTCCGGGCATATCTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIT1 - tRNA isopentenyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312691.1 | 1236 | Intron | NP_001299620.1 | |||
NM_001312692.1 | 1236 | Missense Mutation | CAA,CGA | Q,R 245 | NP_001299621.1 | |
NM_017646.5 | 1236 | Missense Mutation | CAA,CGA | Q,R 327 | NP_060116.2 | |
XM_005270954.2 | 1236 | Missense Mutation | CAA,CGA | Q,R 246 | XP_005271011.1 | |
XM_006710706.1 | 1236 | Missense Mutation | CAA,CGA | Q,R 186 | XP_006710769.1 |