Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAAGAGAAGACCTTACAACAGG[A/T]GGACAGGATGCTGGCTGAGGTTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
METTL13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METTL13 - methyltransferase like 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007239.1 | 430 | Missense Mutation | GAG,GTG | E,V 139 | NP_001007240.1 | |
NM_014955.2 | 430 | Missense Mutation | GAG,GTG | E,V 53 | NP_055770.1 | |
NM_015935.4 | 430 | Missense Mutation | GAG,GTG | E,V 139 | NP_057019.3 | |
XM_017001455.1 | 430 | Missense Mutation | GAG,GTG | E,V 53 | XP_016856944.1 | |
XM_017001456.1 | 430 | Missense Mutation | GAG,GTG | E,V 53 | XP_016856945.1 | |
XM_017001457.1 | 430 | Missense Mutation | GAG,GTG | E,V 53 | XP_016856946.1 | |
XM_017001458.1 | 430 | Missense Mutation | GAG,GTG | E,V 139 | XP_016856947.1 | |
XM_017001459.1 | 430 | Missense Mutation | GAG,GTG | E,V 53 | XP_016856948.1 |