Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTTTGCAGGCCCCTTGGACCGC[C/T]GGCCAGATATTCAGGCGGATCAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604973 | ||||||||||||||||||||
Literature Links: |
CD164L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD164L2 - CD164 molecule like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005245868.3 | 220 | Silent Mutation | CCA,CCG | P,P 48 | XP_005245925.2 | |
XM_011541441.1 | 220 | Silent Mutation | CCA,CCG | P,P 48 | XP_011539743.1 |
FCN3 - ficolin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |