Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAGCAGGGCACGCAGTAGTGAGC[A/T]CCCTTGTCGGGCACAAAAGAACGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602790 MIM: 605596 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FHL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FHL3 - four and a half LIM domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243878.1 | 441 | Silent Mutation | GGA,GGT | G,G 38 | NP_001230807.1 | |
NM_004468.4 | 441 | Silent Mutation | GGA,GGT | G,G 146 | NP_004459.2 | |
XM_017000675.1 | 441 | Intron | XP_016856164.1 |
SF3A3 - splicing factor 3a subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |