Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCCTGAGCTGGACACCTGGAAC[C/T]GTGTTGAACTTGTGCTTCTTGAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606822 MIM: 613170 | ||||||||||||||||||||
Literature Links: |
POMGNT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POMGNT1 - protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243766.1 | 1610 | Silent Mutation | ACA,ACG | T,T 524 | NP_001230695.1 | |
NM_001290129.1 | 1610 | Silent Mutation | ACA,ACG | T,T 502 | NP_001277058.1 | |
NM_001290130.1 | 1610 | Silent Mutation | ACA,ACG | T,T 381 | NP_001277059.1 | |
NM_017739.3 | 1610 | Silent Mutation | ACA,ACG | T,T 524 | NP_060209.3 | |
XM_005271010.1 | 1610 | Silent Mutation | ACA,ACG | T,T 524 | XP_005271067.1 | |
XM_006710755.1 | 1610 | Silent Mutation | ACA,ACG | T,T 524 | XP_006710818.1 | |
XM_006710756.1 | 1610 | Silent Mutation | ACA,ACG | T,T 524 | XP_006710819.1 | |
XM_011541760.2 | 1610 | Silent Mutation | ACA,ACG | T,T 502 | XP_011540062.1 | |
XM_017001690.1 | 1610 | Silent Mutation | ACA,ACG | T,T 524 | XP_016857179.1 |
TSPAN1 - tetraspanin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005727.3 | 1610 | Intron | NP_005718.2 | |||
XM_011540460.2 | 1610 | Intron | XP_011538762.1 |