Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTACGAGGATGGCACGGAGCTGA[C/T]GGAAGATTACTTCCCCAGTGTTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616690 MIM: 601883 | ||||||||||||||||||||
Literature Links: |
CEP104 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEP104 - centrosomal protein 104 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DFFB - DNA fragmentation factor subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282669.1 | 504 | Missense Mutation | ACG,ATG | T,M 56 | NP_001269598.1 | |
NM_001320132.1 | 504 | UTR 5 | NP_001307061.1 | |||
NM_001320136.1 | 504 | UTR 5 | NP_001307065.1 | |||
NM_004402.3 | 504 | Missense Mutation | ACG,ATG | T,M 56 | NP_004393.1 | |
XM_011540865.2 | 504 | Missense Mutation | ACG,ATG | T,M 56 | XP_011539167.1 | |
XM_017000498.1 | 504 | Missense Mutation | ACG,ATG | T,M 56 | XP_016855987.1 | |
XM_017000499.1 | 504 | Missense Mutation | ACG,ATG | T,M 56 | XP_016855988.1 | |
XM_017000500.1 | 504 | Missense Mutation | ACG,ATG | T,M 56 | XP_016855989.1 |